Order Entry
Belgium
ContactUsLinkComponent
Chloramine-T (sodium salt) trihydrate 97+%
Irritant
Health
Corrosive
Chloramine-T (sodium salt) trihydrate 97+%
Certificates
undefined
Irritant
Health
Corrosive
Chloramine-T (sodium salt) trihydrate 97+%
Certificates

Specification Test Results

  • Appearance:
    White to almost white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration Iodometric:
    97.0 to 103.0 %
  • Appearance of solution:
    (5% in water) Clear solution
  • pH:
    9.0 to 11.0
  • Color of solution:
    ≤25 APHA (5 % in water)

Specification Test Results

Related Information

Frequently Bought Together