Order Entry
Belgium
ContactUsLinkComponent
2'-Hydroxy-5'-methoxyacetophenone 99%
Irritant
2'-Hydroxy-5'-methoxyacetophenone 99%
Certificates
undefined
Irritant
2'-Hydroxy-5'-methoxyacetophenone 99%

Specification Test Results

  • Appearance:
    Yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    48 to 51 °C
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together