
Methyl orange
Supplier: Roth Carl
Certificates
About this item
CAS Number:547-58-0
:
Formula:C₁₄H₁₄N₃NaO₃S
MDL Number:MFCD00007502
:327.34 g/mol
Merck Index:13,06127
Boiling Pt:∼90 °C (1013 hPa)
Melting Pt:> 300 °C
Density:∼1 g/cm³ (20 °C)
Flash Pt:38 °C
Storage Temperature:Ambient
EINECS:208-925-3
UN:3143
ADR:6.1,III
:GHS06
Frequently Bought Together