Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Xylan (from corn core)

 :   TCI
Xylan (from corn core)
 :  TCI
 
Xylan (from corn core)

 

 
  •  :9014-63-5
  •  :MFCD00082148
  •  :232-760-6
 

 

  • Appearance:
    White to Yellow to Orange powder to crystal
  • Xylose:
    min. 75.0 % (after hydrolysis)
  • Drying loss:
    max. 15.0 %
Product Family Options

 

  • Pk
     
     
  • 25 g
       
       
    1 * 25 g
    1 * 25 g - 46,20 €
    46,20 €
    1 * 25 g
       46,20 €

  • 100 g
       
       
    1 * 100 g
    1 * 100 g - 128,00 €
    128,00 €
    1 * 100 g
       128,00 €