4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98.0% (by GC)
Catalog # TCIAT3747-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 96.0 %
- Melting point:175.0 to 179.0 deg-C
Specification Test Results
About this item
Synonyms: (9H-Carbazol-4-yl)boronic acid Pinacol ester, 4-(9H-Carbazol-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
:
:293.17 g/mol
:GHS07
CAS Number:1255309-13-7
Formula:C₁₈H₂₀BNO₂
MDL Number:MFCD16996079