Order Entry
Belgium
ContactUsLinkComponent
Tissue-clearing reagent, CUBIC-R+ for animals
Tissue-clearing reagent, CUBIC-R+ for animals
Catalog # TCIAT3741-25ML
Supplier:  TCI
CAS Number:  
Tissue-clearing reagent, CUBIC-R+ for animals
Catalog # TCIAT3741-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Description:
    Tissue-Clearing reagent, CUBIC-R+ for Animals

Specifications

About this item

Specifications
Appearance: Colorless to Light orange to Yellow clear liquid
Functionality test: to pass test (Remaining fluorescence strength)