Order Entry
Belgium
ContactUsLinkComponent
Pararosaniline solution, Sigma-Aldrich®
Health
Corrosive
Irritant
Pararosaniline solution, Sigma-Aldrich®
Catalog # 1804-15ML
CAS Number:  
Health
Corrosive
Irritant
Pararosaniline solution, Sigma-Aldrich®
Catalog # 1804-15ML
CAS Number:  

Specifications

  • Pk:
    15 mL
  • Seq:
    2295

Specifications

About this item

 :
 :305.38 g/mol
ADR:8,II
 :GHS08
 :GHS05
 :GHS07
Storage Temperature:Ambient
 :1
CAS Number:467-62-9
 :25620-78-4
Formula:C₁₉H₁₉N₃O
MDL Number:MFCD00036222