9-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbazole ≥98.0% (by GC)
Leverancier: TCI
Certificaten
Over dit artikel
Synoniemen: 9-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-9H-carbazole, 2-[3-(9H-Carbazol-9-yl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-(9H-Carbazol-9-yl)phenylboronic acid Pinacol ester
CAS-nummer:870119-58-7
:
Formule:C₂₄H₂₄BNO₂
MDL-nummer:MFCD22415352
:369.27 g/mol
:GHS07
Specificaties
- Environmentally Preferable:
Specificaties
Specificatie testresultaten
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 96.0 %
- Melting point:128.0 to 132.0 deg-C
Specificatie testresultaten
Frequently Bought Together