1-(Trimethoxysilyl)naphthalene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: Trimethoxy(1-naphthyl)silane
CAS Number:18052-76-1
:
Formula:C₁₃H₁₆O₃Si
MDL Number:MFCD00094090
Boiling Pt:148°C
:248.35 g/mol
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 98.0 %
- Melting point:37.0 to 41.0 deg-C
Specification Test Results
Frequently Bought Together