Order Entry
Belgium
ContactUsLinkComponent
1-(p-Toluenesulphonyl)-1,2,4-triazole ≥98.0% (by HPLC)
Irritant
1-(p-Toluenesulphonyl)-1,2,4-triazole ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
undefined
Irritant
1-(p-Toluenesulphonyl)-1,2,4-triazole ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Melting point:
    105.0 to 109.0 deg-C

Specification Test Results

Frequently Bought Together