Order Entry
Belgium
ContactUsLinkComponent
3,3,3-Trifluoro-2-hydroxy-2-(trifluoromethyl)propionic acid ≥98.0% (by titrimetric analysis)
Corrosive
3,3,3-Trifluoro-2-hydroxy-2-(trifluoromethyl)propionic acid ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
Corrosive
3,3,3-Trifluoro-2-hydroxy-2-(trifluoromethyl)propionic acid ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystaline
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    82.0 to 86.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together