(R)-3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAT1900-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Optical purity (GC):min. 98.0 ee%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:108.0 to 112.0 deg-C
Specification Test Results
About this item
:
:158.08 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05
CAS Number:44864-47-3
Formula:C₄H₅F₃O₃
MDL Number:MFCD03095400