Tetrabutylphosphonium bis(1,3-dithiole-2-thione-4,5-dithiolato)nickel(III) complex ≥98.0% (by titrimetric analysis)
Catalog # TCIAT1416-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Dark green to Dark blue to Black powder to crystal
- Purity (Chelometric Titration):min. 98.0 %
Specification Test Results
About this item
:
:710.80 g/mol
:GHS08
CAS Number:105029-70-7
Formula:C₂₂H₃₆NiPS₁₀
MDL Number:MFCD00191494