Tetrabutylammonium salicylate ≥98.0% (by HPLC)
Catalog # TCIAT1276-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:90.0 to 94.0 deg-C
Specification Test Results
About this item
Synonyms: Tetrabutylammonium 2-hydroxybenzoate
:
:379.59 g/mol
CAS Number:22307-72-8
Formula:C₂₃H₄₁NO₃
MDL Number:MFCD00145388