Order Entry
Belgium
ContactUsLinkComponent
Dextran solution 20% (from Leuconostoc mesenteroides), autoclaved, Sigma-Aldrich®
Dextran solution 20% (from Leuconostoc mesenteroides), autoclaved, Sigma-Aldrich®
Certificates
Dextran solution 20% (from Leuconostoc mesenteroides), autoclaved, Sigma-Aldrich®

Specification Test Results

  • Appearance (Turbidity):
    Clear to Slightly Hazy
  • Appearance (Colour):
    Colorless
  • Appearance (Form):
    Liquid
  • Density:
    1.00 - 1.20 g/ml

Specification Test Results

Frequently Bought Together