Sodium-3-[ethyl(m-tolyl)amino]-2-hydroxy-1-propanesulphonate hydrate ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:82692-93-1
:
Formula:C₁₂H₁₉NO₄S
MDL Number:MFCD00040641
:295.33 g/mol
EINECS:420-430-6
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 97.0 % (calcd.on anh.substance)
- Water:8.0 to 11.5 %
Specification Test Results
Frequently Bought Together