Order Entry
Belgium
ContactUsLinkComponent
Sodium-1,2,4-triazolide ≥96.0% (by GC, titration analysis)
Irritant
Sodium-1,2,4-triazolide ≥96.0% (by GC, titration analysis)
Catalog # TCIAS0522-25G
Supplier:  TCI
CAS Number:  
Irritant
Sodium-1,2,4-triazolide ≥96.0% (by GC, titration analysis)
Catalog # TCIAS0522-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 96.0 %
  • Purity (Nonaqueous Titration):
    min. 96.0 %

Specification Test Results

About this item

 :
 :91.05 g/mol
Melting Pt:327°C
EINECS:255-280-9
 :GHS07
CAS Number:41253-21-8
Formula:C₂H₃N₃
MDL Number:MFCD00044437