9-Phenyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
Synonyms: 9-Phenylcarbazole-3-boronic acid Pinacol ester, 4,4,5,5-Tetramethyl-2-(9-phenylcarbazol-3-yl)-1,3,2-dioxaborolane, 9-Phenyl-9H-carbazol-3-yl-3-boronic acid pinacol ester, 9-Phenyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
CAS Number:1126522-69-7
:
Formula:C₂₄H₂₄BNO₂
MDL Number:MFCD16621140
:369.27 g/mol
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:162.0 to 166.0 deg-C
Specification Test Results
Frequently Bought Together