Order Entry
Belgium
ContactUsLinkComponent
 
 
Palladium(II) acetate ≥98.0% (by titrimetric analysis), purified, purified by sublimation
  TCIAP2161-1G
 :  TCI
 :  
undefined
 
Palladium(II) acetate ≥98.0% (by titrimetric analysis), purified, purified by sublimation
  TCIAP2161-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (Chelometric Titration):
    min. 98.0 %
  • Elemental analysis (Nitrogen):
    max. 0.30 %

 

 

 :
 :224.51 g/mol
 :14,06991
 :222-164-4
 :GHS05
 :3375-31-3
 :C₄H₆O₄Pd
 :MFCD00012453