N-(Pyrazinylcarbonyl)-L-phenylalanine ≥98.0% (by HPLC)
Catalog # TCIAP2068-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 95.0 %
- Melting point:167.0 to 171.0 deg-C
- Specific rotation [a]20/D:+9.0 to +12.0 deg (C=1,MeOH)
Specification Test Results
About this item
:
:271.28 g/mol
CAS Number:114457-94-2
Formula:C₁₄H₁₃N₃O₃
MDL Number:MFCD09800536