
N-Phthaloyl-L-phenylalanine ≥98.0% (by HPLC, titration analysis)
TCIAP1227-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:183.0 to 186.0 deg-C
- Specific rotation [a]20/D:-210.0 to -219.0 deg (C=2, EtOH)
:
:295.29 g/mol
:GHS07
:5123-55-7
:C₁₇H₁₃NO₄
:MFCD00069738