Order Entry
Belgium
ContactUsLinkComponent
Morin hydrate ≥90.0% (by HPLC)
Environment
Morin hydrate ≥90.0% (by HPLC)
Supplier:  TCI
Certificates
Environment
Morin hydrate ≥90.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (HPLC):
    min. 90.0 area%
  • Water:
    max. 12.0 %

Specification Test Results

Related Information

Frequently Bought Together