DL-Pantolactone ≥95.0% (by GC)
: TCI
: Dihydro-3-hydroxy-4,4-dimethyl-2(3H)-furanone, DL-Pantoyl Lactone, 3-Hydroxy-4,4-dimethyldihydrofuran-2(3H)-one, DL-2-Hydroxy-3,3-dimethyl-gamma-butyrolactone
:79-50-5
:
:C₆H₁₀O₃
:MFCD00064333
:121°C
:78°C
:130.14 g/mol
:201-210-7
- Environmentally Preferable:
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
Frequently Bought Together