L-Ornithine L-Aspartate ≥96.0% (by titrimetric analysis)
TCIAO0440-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 96.0 %
- Specific rotation [a]20/D:+26.0 to +29.0 deg (C=1, 6mol/L HCl)
:
:265.27 g/mol
:14,06874
:Refrigerator
:221-772-7
:3230-94-2
:C₉H₁₉N₃O₆
:MFCD00058084