Methyl-2-oxo-1-pyrrolidineacetate ≥97.0% (by GC)
: TCI
: 2-Oxo-1-pyrrolidineacetic Acid Methyl Ester, 2-Pyrrolidinone-1-acetic Acid Methyl Ester
:59776-88-4
:
:C₇H₁₁NO₃
:MFCD00003198
:157.17 g/mol
:261-926-0
- Environmentally Preferable:
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
Frequently Bought Together