1-(2-Nitrophenyl)piperazine ≥98.0% (by GC)
: TCI
:59084-06-9
:
:C₁₀H₁₃N₃O₂
:MFCD00040728
:126°C
:207.23 g/mol
:261-593-1
:GHS07
- Environmentally Preferable:
- Appearance:Orange to Red clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together