Order Entry
Belgium
ContactUsLinkComponent
 
3-(4-Methoxyphenyl)-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)
 :  TCI
 
3-(4-Methoxyphenyl)-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    159.0 to 163.0 deg-C

 

Frequently Bought Together