
2-(4-Methyl-5-thiazolyl)ethyl hexanoate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:94159-32-7
:
Formula:C₁₂H₁₉NO₂S
MDL Number:MFCD09032914
:241.35 g/mol
Boiling Pt:152°C
EINECS:303-210-3
:GHS07
Specification Test Results
- Appearance:White to Almost white clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together