m-PEG12-NHS ester
Catalog # TCIAM2188-25MG
Supplier: TCI
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
Specification Test Results
- Appearance:White to Almost white cloudy liquid
- Elemental analysis (Nitrogen):1.80 to 2.20 %
Specification Test Results
About this item
:
:685.755 g/mol
Storage Temperature:Freezer
CAS Number:756525-94-7
Formula:C₃₀H₅₅NO₁₆
MDL Number:MFCD13184970