m-PEG8-NHS ester
25 mg
Catalog # TCIAM2187-25MG
Supplier: TCI
Jump to:
Product Details & Documents
Synonyms: N-Succinimidyl 4,7,10,13,16,19,22,25-Octaoxahexacosanoate, 2,5-Dioxopyrrolidin-1-yl 2,5,8,11,14,17,20,23-octaoxahexacosan-26-oate, Methyl-PEG8-NHS Ester, mPEG8-NHS Ester, 4,7,10,13,16,19,22,25-Octaoxahexacosanoic Acid N-Succinimidyl Ester
- CAS Number:756525-90-3
- Formula:C₂₂H₃₉NO₁₂
- MDL Number:MFCD13184969
- Molecular Weight:509.54 g/mol
- Storage Temperature:Freezer
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Elemental analysis (Nitrogen):2.50 to 3.00 %
Specifications
- Pk:25 mg



