m-PEG4-NHS ester
25 mg
Catalog # TCIAM2186-25MG
Supplier: TCI
Jump to:
Product Details & Documents
Synonyms: mPEG4-NHS Ester, Methyl-PEG4-NHS Ester, N-Succinimidyl 4,7,10,13-Tetraoxatetradecanoate, 2,5-Dioxopyrrolidin-1-yl 2,5,8,11-tetraoxatetradecan-14-oate, 4,7,10,13-Tetraoxatetradecanoic acid N-succinimidyl ester
- CAS Number:622405-78-1
- Formula:C₁₄H₂₃NO₈
- MDL Number:MFCD11041112
- Molecular Weight:333.34 g/mol
- Storage Temperature:Freezer
Documents
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Elemental analysis (Nitrogen):3.80 to 4.60 %
Specifications
- Pk:25 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



