5H-5-Methyl-6,7-dihydrocyclopentapyrazine ≥97.0% (by GC, titration analysis)
Catalog # TCIAM1086-5ML
Supplier: TCI
CAS Number:
Specifications
- Pk:5 mL
- Seq:2210
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
About this item
Synonyms: 5-Methyl-6,7-dihydro-5H-cyclopenta[b]pyrazine, 5-Methyl-2,3-cyclopentenopyrazine
:
:134.18 g/mol
Boiling Pt:80°C
Flash Pt:79°C
EINECS:245-864-1
:GHS07
CAS Number:23747-48-0
Formula:C₈H₁₀N₂
MDL Number:MFCD00040997