Order Entry
Belgium
ContactUsLinkComponent
3-Methyl-1-p-tolyl-5-pyrazolone ≥98.0% (by HPLC, titration analysis)
3-Methyl-1-p-tolyl-5-pyrazolone ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAM0554-25G
Supplier:  TCI
CAS Number:  
3-Methyl-1-p-tolyl-5-pyrazolone ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAM0554-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    136.0 to 142.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :188.23 g/mol
EINECS:201-708-4
CAS Number:86-92-0
Formula:C₁₁H₁₂N₂O
MDL Number:MFCD00035708