Leuprorelin acetate
Catalog # TCIAL0249-25MG
Supplier: TCI
CAS Number:
Specifications
- Pk:25 mg
- CAS No.:74381-53-6
- Storage conditions:Freezer
- Form:White to light yellow powder to crystal
- MDL No.:MFCD00167544
- Protein/peptide name:Leuprorelin acetate
- Purity:Min. 97,0% (HPLC)
- Molecular weight:1269,47 g/mol
- Molecular formula:C₅₉H₈₄N₁₆O₁₂
Specifications
About this item
- Specific rotation [a]20/D: –31.0 to –35.0° (C=1,AcOH (1%))
- Merck index: 14,05457
Product Categories: Bioscience, Antitumor Ingredients, Oligopeptides
Other Product Names / Synonyms: Leuprolide Acetate