D-Lactose monohydrate ≥98.0% (by HPLC)
: TCI
:64044-51-5
:63-42-3
:C₁₂H₂₂O₁₁·1H₂O
:MFCD00150747
:360.32 g/mol
:14,05343
:238-691-8
- Appearance:White powder to crystal
- Purity (HPLC) (CAD):min. 98.0 area%
- Specific rotation [a]20/D:+54.4 to +59.9 deg (C=10, H2O) (calcd.on anh.substance)
- Solubility in Water:within almost transparency
- Water:4.5 to 5.5 %
Frequently Bought Together