9-Hexyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
Synonyms: 2-(9-Hexyl-9H-carbazol-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 9-Hexyl-9H-carbazole-2-boronic acid Pinacol ester
CAS Number:1339953-35-3
:
Formula:C₂₄H₃₂BNO₂
:377.34 g/mol
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Melting point:83.0 to 87.0 deg-C
Specification Test Results
Frequently Bought Together