
(3-Hydroxy-4-methoxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1131-94-8
:
Formula:(C₆H₃(OH)(OCH₃))CH₂COOH
MDL Number:MFCD00016829
:182.18 g/mol
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:129.0 to 133.0 deg-C
Specification Test Results
Frequently Bought Together