3-(4-Hydroxy-3-methoxyphenyl)propionic acid ≥98.0% (by GC)
: TCI
:1135-23-5
:
:C₁₀H₁₂O₄
:MFCD00016558
:196.20 g/mol
:214-489-5
:GHS07
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 96.0 %
- Melting point:89.0 to 93.0 deg-C
Frequently Bought Together