



Hydrobromic acid 10 - 20% in ethanol for esterification
Catalog # TCIAH0959-25ML
Supplier: TCI
CAS Number:
Specifications
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Concentration (Neutralization titration):10.0 to 20.0 w/w%
Specification Test Results
About this item
:
:80.91 g/mol
Merck Index:14,04778
Storage Temperature:Refrigerator
EINECS:233-113-0
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:10035-10-6
Formula:HBr
MDL Number:MFCD00011323