
Ethyl-2-hydroxy-3-butynoate ≥95.0% (by GC)
Catalog # TCIAH0823-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Yellow to Green clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
About this item
:
:128.13 g/mol
Boiling Pt:78°C
:GHS07
Storage Temperature:Freezer
CAS Number:18418-08-1
Formula:C₆H₈O₃
MDL Number:MFCD00191654