3-β-Hydroxy-δ-5-cholenic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:5255-17-4
:
Formula:C₂₄H₃₈O₃
MDL Number:MFCD00069468
:374.57 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Neutralization titration):min. 97.0 %
- Specific rotation [a]20/D:-38.0 to -41.0 deg (C=1, Dioxane)
- Solubility in hot Dioxane:almost transparency
Specification Test Results
Frequently Bought Together