8-Hydroxy-7-iodoquinoline-5-sulphonic acid ≥98.0% (by HPLC, titration analysis)
TCIAH0254-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Solubility in hot Water:very faint turbidity
:
:351.11 g/mol
:14,04826
:208-938-4
:3261
:8,III
:GHS05
:547-91-1
:C₉H₆INO₄S
:MFCD00006793