D-(+)-Glucosamine hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:66-84-2
:
Formula:C₆H₁₃NO₅
MDL Number:MFCD00135831
:215.63 g/mol
EINECS:200-638-1
Specification Test Results
- Appearance:White powder to crystaline
- Purity (Argentmetric Titration):min. 98.0 %
- Specific rotation [a]20/D:+70.0 to +74.0 deg (C=1, H2O) (after 20hours)
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together