
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid (trans-) ≥98.0% (by GC, titration analysis), purified by sublimation
Supplier: TCI
Certificates
About this item
CAS Number:537-98-4
:1135-24-6
Formula:C₁₀H₁₀O₄
MDL Number:MFCD00004400
:194.18 g/mol
EINECS:208-679-7
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:172.0 to 176.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together