Order Entry
Belgium
ContactUsLinkComponent
(Fmoc-amino)-PEG6-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Irritant
(Fmoc-amino)-PEG6-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAF1183-250MG
Supplier:  TCI
CAS Number:  
Irritant
(Fmoc-amino)-PEG6-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAF1183-250MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :575.647 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:882847-34-9
Formula:C₃₀H₄₁NO₁₀
MDL Number:MFCD06656474