N-Formyl-L-Methionyl-L-Leucyl-L-Phenylalanine
Supplier: TCI
Certificates
About this item
- Specific rotation [a]20/D: –8,0 to –10,0° (C=1, acetic acid)
Specifications
- CAS No.:59880-97-6
- Storage conditions:Freezer
- Form:White to almost white powder to crystal
- MDL No.:MFCD00036780
- Protein synonyms:N-Formyl-Met-Leu-Phe|fMLP|fMLF
- Protein/peptide name:N-Formyl-L-methionyl-L-leucyl-L-phenylalanine
- Purity:98,0% (HPLC)
- Molecular weight:437,56 g/mol
- Molecular formula:C₂₁H₃₁N₃O₅S
Specifications
Frequently Bought Together