2-Fluoro-6-(trifluoromethyl)benzaldehyde ≥98.0% (by GC)
Leverancier: TCI
Certificaten
Over dit artikel
CAS-nummer:60611-24-7
:
Formule:C₈H₄F₄O
MDL-nummer:MFCD00061273
:192.11 g/mol
Kokend Pt:67°C
Flits Pt:82°C
:GHS07
Specificaties
- Environmentally Preferable:
Specificaties
Specificatie testresultaten
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specificatie testresultaten
Frequently Bought Together