Order Entry
Belgium
Orders LinkContactUsLinkComponent
Finasteride ≥98.0% (by GC)

Finasteride ≥98.0% (by GC)

Irritant
Supplier:   TCI
Finasteride ≥98.0% (by GC)
Supplier:  TCI
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  N-tert-Butyl-3-oxo-4-aza-5alpha-androst-1-en-17beta-carboxamide
  • CAS Number:98319-26-7
  • Formula:C₂₃H₃₆N₂O₂
  • MDL Number:MFCD00869737
  • Molecular Weight:372.55 g/mol
  • Merck Index:14,04082
  • Melting Pt:253°C
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 34,10 €
    34,10 €
    1 * 200 mg
       34,10 €

  • 1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 128,00 €
    128,00 €
    1 * 1 g
       128,00 €