5-Fluoro-o-tolylisothiocyanate ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:175205-39-7
:
Formula:C₈H₆FNS
MDL Number:MFCD00046800
:167.20 g/mol
:GHS07
Specification Test Results
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (GC):min. 98.0 %
- Purity (Butylamine Method):min. 98.0 %
Specification Test Results
Related Information
Frequently Bought Together