N-Formyl-L-phenylalanine ≥98.0% (by titrimetric analysis)
TCIAF0127-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:163.0 to 168.0 deg-C
- Specific rotation [a]20/D:+73.0 to +75.0 deg (C=4, EtOH)
- Solubility in EtOH:almost transparency
:
:193.20 g/mol
:236-166-8
:13200-85-6
:C₁₀H₁₁NO₃
:MFCD00021030